Learn More
Sonoco™ ThermoSafe LabMailer Specimen Tube Mailers
Product Code. p-150224
Change view
Click to view available options
Capacity:
7, 10, 15 or 25 mL
Quantity:
5 Tubes
Unit Size:
Pack of 100
Product Code. Capacity Quantity unitSize
17808703 7, 10, 15 or 25 mL 5 Tubes Pack of 100
1 options
Product Code. 17808703
Brand: Sonoco™ 409
This item requires a price update prior to being ordered.
Request a Price for this item here.
Shock-absorbing polystyrene foam mailers accept sample tubes with rubber or screw caps
  • Two-piece assembly with snap-lock fasteners holds contents in place
  • Ideal for mailing or transporting nonhazardous blood, urine, or other lab samples.
†Includes slotted area for up to four slides.
TRUSTED_SUSTAINABILITY
Specifications
Quantity 5 Tubes
Applications Ideal for mailing or transporting nonhazardous blood, urine, or other lab samples
Dimensions (L x W x H) Exterior 7.875 x 5.125 x 1.5 in.
Width (Metric) Interior 16 mm
Diameter (Metric) Interior 170 mm
Description Foam only shipper. Must purchase mailing sleeve model 410, Holds 5 Tubes
Capacity 7, 10, 15 or 25 mL
Material Polystyrene Foam