Learn More
Cole-Parmer Thermoplastic Vulcanizate (TPV) Pharma Grade Tubing
Product Code. 30054338 Shop All Cole-Parmer Products
Change view
Click to view available options
Diameter (English) Inner:
1/16 in.
1/2 in.
1/4 in.
1/8 in.
3/16 in.
3/8 in.
Diameter (English) Outer:
1/2 in.
1/4 in.
3/16 in.
3/4 in.
3/8 in.
5/16 in.
5/8 in.
7/16 in.
Unit Size:
15m
9 product options available for selection
Product selection table with 9 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Diameter (English) Inner Diameter (English) Outer Unit Size
30054338 3/16 in. 7/16 in. 15m
30053527 1/2 in. 3/4 in. 15m
30053912 1/4 in. 1/2 in. 15m
30054171 1/16 in. 3/16 in. 15m
30053517 1/4 in. 7/16 in. 15m
30053451 1/4 in. 3/8 in. 15m
30053658 3/16 in. 5/16 in. 15m
30054210 1/8 in. 1/4 in. 15m
30053991 3/8 in. 5/8 in. 15m
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 9 options available.
9 options
Product Code. 30054338 Supplier Cole-Parmer Supplier No. 5011987
  • ONLINE EXCLUSIVE PRICE
    3220.00 NOK
    3016.39 NOK / 15m
    Save 203.61 NOK
    6% Off
Estimated Shipment: 15-02-2027
Offers great chemical and kink resistance for e in., cellent pump performance
Biocompatible thermoplastic tubing offers good chemical resistance. Does not contain BPA, phthalates, PVC, or pero in., ides. Sterilize by autoclave, gamma irradiation, and chemical sterilants. Complies with U.S. Pharmacopeia 43, National Formulary 38 (USP), General Chapter 88 Class VI, Biological reactivity Tests, In Vivo (2020). Ideal for general-purpose, laboratory, and single-use applications. Free technical application support available to assist with tubing selection
Specifications
Certifications/Compliance FDA;Non-Phthalate;USP;RoHS
Diameter (English) Inner 3/16 in.
Diameter (English) Outer 7/16 in.
Overall Length (English) 50 ft.