Learn More
Quickfit™ Thermoplastic Polyester Tubing Clamps
Product Code. 12704406
Change view
Click to view available options
For Use With:
Up to 10mm O.D. tubing
Up to 14mm O.D. tubing
Up to 4.5mm O.D. tubing
Up to 6mm O.D. tubing
Unit Size:
Pack of 5
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. For Use With Unit Size
12704406 Up to 4.5mm O.D. tubing Pack of 5
12316357 Up to 10mm O.D. tubing Pack of 5
11961528 Up to 14mm O.D. tubing Pack of 5
11332025 Up to 6mm O.D. tubing Pack of 5
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Product Code. 12704406 Supplier Quickfit Supplier No. KT4.5
  • ONLINE EXCLUSIVE PRICE
    358.60 NOK
    282.29 NOK / Pack of 5
    Save 76.31 NOK
    21% Off
Estimated Shipment: 01-12-2026
For use with flexible rubber and plastic tubing
  • Provides fine flow control of liquids and gases
  • Easy one handed adjustment: just roll serated wheel along sloped floor
  • Manufactured from thermoplastic polyester
Specifications
Material Thermoplastic Polyester
For Use With Up to 4.5mm O.D. tubing
For Use With (Application) Used with flexible Rubber and Plastic tubing
Product Type Tubing Clamp