Learn More
Thermometer Adapter for ML1 Microscale kit
Product Code. 15788037
Change view
Click to view available options
Product Type:
thermometer Adapter for ML1 Microscale Kit
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Product Type Unit Size
15788037 thermometer Adapter for ML1 Microscale Kit Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 15788037 Supplier Quickfit Supplier No. ML/ST
  • ONLINE EXCLUSIVE PRICE
    243.60 NOK
    210.23 NOK / Pack of 1
    Save 33.37 NOK
    13% Off
Estimated Shipment: 10-02-2027
Designed for organic chemistry experiments.
  • Designed for organic chemistry experiments using milligram quantities of reagents
Specifications
For Use With (Application) For organic chemistry experiments
Product Type thermometer Adapter for ML1 Microscale Kit