Learn More
Thermo Scientific™ Thermo Series Liquid Nitrogen Transfer Vessels
Product Code. 10480394 Shop All Thermo Scientific Products
Change view
Click to view available options
Capacity (Metric):
5 L
10 L
20 L
32 L
Unit Size:
Each
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Capacity (Metric) Unit Size
10480394 5 L Each
10318543 10 L Each
10398293 20 L Each
11587304 32 L Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Product Code. 10480394 Supplier Thermo Scientific™ Supplier No. TY509X1
  • ONLINE EXCLUSIVE PRICE
    19591.76 NOK
    17665.16 NOK / Pack of 1
    Save 1926.60 NOK
    9% Off
 Restricted Item
Estimated Shipment: 29-01-2027
Store and dispense small amounts of liquid nitrogen with our economical and durable liquid nitrogen transfer vessels.
Store and dispense small amounts of liquid nitrogen with our economical and durable Thermo Scientific™ Thermo Series Liquid Nitrogen Transfer Vessels. The Thermo Series includes six models with capacities from 5 to 32 L. A full range of handling and dispensing accessories as well as transport carts are also available.
Features:
  • Designed to bring liquid nitrogen to other cryovessels
  • Lightweight aluminum dewars assure low-static evaporation rates
  • Optional withdrawal device, 12mL dipper and wheeled accessory cart
  • Compact Thermo 5 and 10 dewars feature a convenient pail-style handle for pouring and for use in applications where only small quantities of liquid nitrogen are needed
  • Thermo 10, 20, and 30 can be fitted with self-pressurized withdrawal accessory devices to easily dispense LN2 without pouring
Note:
Stated static evaporation rates are based on new container performance with no lid openings and no product loads. Actual working performance may vary with individual applications.
TRUSTED_SUSTAINABILITY
Specifications
Type LN2 Transfer Vessel
Capacity (Metric) 5 L
Height (English) Exterior 18.2 in.
Height (Metric) Exterior 46.2 cm
Diameter (English) 8.8 in.
Diameter (English) Inside Neck 2.2 in.
Diameter (Metric) 22.4 cm
Diameter (Metric) Inside Neck 5.6 cm
Shipping Weight (English) 12 lb.
Shipping Weight (Metric) 5.4 kg
Weight (English) 8 lb.
Weight (Metric) 3.6 kg