Learn More
Thermo Scientific™ Ultrafast UFC-1 Phase GC Columns
Product Code. p-3666090 Shop All Thermo Scientific Products
Click to view available options
Length (Metric):
10 m
Film Thickness:
0.4 μm
Unit Size:
Each
Product Code. 11531004
Brand: Thermo Scientific™ UFMC00502010006
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
Short, narrow-bore columns for use with the Thermo Scientific TRACE GC Ultra Chromatograph
  • Reduces analytical run time for increased throughput
  • Long column lifetime
TRUSTED_SUSTAINABILITY
Specifications
Length (Metric) 10 m
Product Line Ultrafast
Film Thickness 0.4 μm