Learn More
Unity Lab Services™ Thermo Scientific™ Ultra-Low Temperature Freezers Validation and Temperature Map Documentation
Product Code. 16494974
Change view
Click to view available options
For Use With (Equipment):
ThermoScientific ULT Freezers
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. For Use With (Equipment) Unit Size
16494974 ThermoScientific ULT Freezers Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 16494974 Supplier Unity Lab Services™ Supplier No. IOPQDOCE89003484
 Restricted Item
Estimated Shipment: 30-12-2026
As an original equipment manufacturer (OEM) service provider for Thermo Fisher Scientific, all protocols designed to optimize efficiency are included with each Thermo Scientific Ultra-Low Temperature Freezers Models: Thermo Scientific TDE, Forma FDE, Revco RDE, Isotemp IUE
IQ/OQ/Temperature Mapping documentation that supports regulatory requirements and industry standards for instrument compliance. Our documents are kept upto date by conducting annual reviews. We work directly with our product experts to have the most updated documentation available.
TRUSTED_SUSTAINABILITY
Specifications
For Use With (Equipment) ThermoScientific ULT Freezers
Product Type IQ/OQ/PQ Documentation
Location Contact your local sales representative for a list of standard and extended coverage areas.
Includes IQ/OQ/Temp Map Documentation