Learn More
Thermo Scientific™ Tips for Tecan™ and Axon™ Instruments PatchXpress Workstations
Description
For use with Tecan™ Genesis™, Freedom Evo™, Cavro™, PatchXpress Workstations
Clear pipet tips, Tecan style.
Specifications
Specifications
| Volume (Metric) | 50 μL |
| Filtered | Filtered |
| Sterility | Non-sterile |
| Color | White |
| Autoclavable | Autoclavable |
| Clean Claims | DNase, RNase, and Pyrogen free |
| Format | Rack |
| Tip Style | Automation |
| For Use With (Equipment) | Tecan Genesis, Tecan Freedom EVO, Cavro, patchXpress |
| Material | Polypropylene |