Learn More
Thermo Scientific™ Sorvall™ Legend™ T/RT Centrifuge Tube Adapters for Round Rotor Buckets
Shop All Thermo Scientific Products
Click to view available options
Color:
Green
Light Gray
Olive Brown
Red
White
Yellow
Tube Size:
10/15 mL
15 mL
175/250 mL
50 mL
7 mL
Unit Size:
Each
Description
Adapters sold individually; order correct quantity to equip your rotor.
Specifications
Specifications
| For Use With (Equipment) | LH-4000W and LH-4000 rotor with round bucket |
| Color | White |
| Tube Size | 175/250 mL |