Learn More
Thermo Scientific™ Sorvall™ Legend™ T/RT Centrifuge Tube Adapters for Four-Place Swing Bucket Bioshield Rotor
For use with four-place Bioshield rotor 75-006-435 and with four-place rotor 75-006-445 when equipped with rectangular bucket 75-006-446
1580.00 NOK - 2245.00 NOK
Product Code | Brand | Description | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Description | Price | Quantity & Availability | |||||
10763073
|
Thermo Scientific™
75006452 |
Adapter, 56 x 1.5/2.0 mL Tubes |
2245.00 NOK
2 sets |
In Stock Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10266033
|
Thermo Scientific™
75006454 |
Adapter, 20 x 7 mL Tubes |
1580.00 NOK
Each |
In Stock Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10260262
|
Thermo Scientific™
75006461 |
Adapter, 4 x 50mL Conical Tubes |
1580.00 NOK
Each |
In Stock Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10215703
|
Thermo Scientific™
75006491 |
Adapter, 4 x 45/50 mL Tubes |
1625.00 NOK
Each |
In Stock Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
Adapters sold individually; order correct quantity to equip rotor.