Learn More
Thermo Scientific™ Sorvall™ Legend™ Centrifuge Carts
Product Code. 10381712 Shop All Thermo Scientific Products
Change view
Click to view available options
Width (Metric):
55.1 cm
74.9 cm
Height (Metric):
29.0 cm
59.9 cm
Unit Size:
Each
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Width (Metric) Height (Metric) unitSize
10381712 74.9 cm 29.0 cm Each
10040112 55.1 cm 29.0 cm Each
10710424 55.1 cm 59.9 cm Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Product Code. 10381712 Supplier Thermo Scientific™ Supplier No. 76000076
  • ONLINE EXCLUSIVE PRICE
    26953.77 NOK
    26673.22 NOK / Pack of 1
    Save 280.55 NOK
    1% Off
Estimated Shipment: 09-10-2026
Add to basket
Add to basket
Allow you to operate your centrifuge operation away from, or under, your primary workbench
  • For all Legend T Series centrifuges; sizes as shown in the table
  • Available in bench-high or under-cabinet models
  • With casters
TRUSTED_SUSTAINABILITY
Specifications
For Use With (Equipment) Thermo Scientific™ Sorvall™ Legend™ RT, Mach 1.6R
Length (Metric) 66.5 cm
Height (Metric) 29.0 cm
Width (Metric) 74.9 cm
Product Type Mobile Cart