Learn More
Thermo Scientific™ Sorvall™ HT6 Tube Adapters for 6-Place Swinging Bucket Rotor with Round Buckets
Product Code. 12468682 Shop All Thermo Scientific Products
Change view
Click to view available options
For Use With (Equipment):
Swinging bucket rotor for HT6 centrifuge
Unit Size:
Pack of 6
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. For Use With (Equipment) Unit Size
12468682 Swinging bucket rotor for HT6 centrifuge Pack of 6
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 12468682 Supplier Thermo Scientific Supplier No. 11178308
  • ONLINE EXCLUSIVE PRICE
    10035.00 NOK
    9195.24 NOK / Pack of 6
    Save 839.76 NOK
    8% Off
 Restricted Item
Estimated Shipment: 29-01-2027
For use with Sorvall HT6 centrifuge
Fits in rotor 11178252 when equipped with round buckets 11178135 (sealed) or 11178136 (unsealed).
Order Info
Sold in packs of 6. One pack will fill the rotor.
TRUSTED_SUSTAINABILITY
Specifications
For Use With (Equipment) Swinging bucket rotor for HT6 centrifuge