Learn More
Thermo Scientific™ Sorvall™ Culture Flask Adapters
Shop All Thermo Scientific Products
Click to view available options
Tube Type:
T-25 Flask
T-75 Flask
Unit Size:
Pack of 2
Description
Requires
Rotor 75006445 with round bucket 75006441
Unique angled design ensures tight pelletization, allowing easy removal of supernatant without pellet disruption.
- T-25 adapter holds flask at 25° angle; T-75 at 75°
- Sold in pairs
Specifications
Specifications
| Tube Type | T-75 Flask |
| For Use With (Equipment) | Thermo Scientific™ Sorvall™ Legend™ T/RT rotor with round bucket |
| Angle | 75° |