Learn More
Thermo Scientific™ Sorvall™ Adapters for Stratos™ Four-Place Swinging Bucket Rotor
Shop All Thermo Scientific ProductsDescription
- Accommodate tube sizes from 1.5 to 100mL, including urine tubes
Order Info
Adapters are sold individually; order correct quantity to equip rotor.
Specifications
Specifications
| For Use With (Equipment) | 4 x 180 Swing bucket rotor |
| Color | Red |