Description
- Properly hardened, tempered and passivated
- Electronically polished and cleaned for a high-quality finish
- Patterns, dimensions and finish held to exacting standards, producing the finest instruments available
- Guaranteed to be free from defects in material and workmanship
Specifications
Specifications
| Product Type | Forceps |
| Grade | Premium |
| Teeth Count | 2 x 3 |
| Length (English) | 5.5 in. |
| Length (Metric) | 14.0 cm |