Learn More
Epredia™ Shandon™ Plastic Coverplates
Product Code. p-4012383 Shop All Epredia Products
Change view
Click to view available options
Quantity:
25/Pk.
250/Cs.
Unit Size:
Pack of 25
Pack of 250
Product Code. Quantity unitSize
11927774 250/Cs. Pack of 250
12766807 25/Pk. Pack of 25
2 options
Product Code. 11927774
Brand: Epredia™ 72110013
For use on Shandon Sequenza™ and Cadenza™ Immunostainers
Saves on the use of costly reagents.
  • Device that fits over standard microscope slide allowing specimens to be immunostained with a minimum amount of reagent
  • Frees considerable technician time
  • Eliminates handling of slides between steps
  • Allows sections to be placed virtually anywhere on slide
  • Protects specimen from drying during long incubations
  • Ensures consistency in staining quality
TRUSTED_SUSTAINABILITY
Specifications
Product Type Coverplate
Quantity 250/Cs.