Learn More
Epredia™ Consul-Mount™
Product Code. 12590017 Shop All Epredia Products
Change view
Click to view available options
Product Type:
Cytology Media
Mountant
Unit Size:
Pack of 4
Product Code. Product Type unitSize
12590017 Cytology Media Pack of 4
12658016 Mountant Pack of 4
2 options
Product Code. 12590017
Brand: Epredia™ 9990441
Designed specifically for use with the Epredia™ Consul™ coverslipper
  • Permanent, organic mountant
  • Xylene base
  • Refractive index: 1.495 ±0.005 (dry)
  • Viscosity: Medium (125-200 cps at 24°C)
TRUSTED_SUSTAINABILITY
Specifications
Color Clear
Product Type Cytology Media
Quantity 500 mL
For Use With (Application) For use with Epredia™ Consul™ coverslipper
Form Liquid