Learn More
Descripción
Easily and quickly cap both the lysis and assay tubes in the Thermo Scientific™ SureTect™ PCR Assay workflow using the Thermo Scientific ™PCR Capping Tool.
Easily and quickly cap both the lysis and PCR tubes in the Thermo Scientific™ SureTect™ PCR Assay workflow using the Thermo Scientific™ PCR Capping Tool.
Designed specifically for use with the SureTect assays, the Thermo Scientific™ Capping Tool is one in a suite of tools enabling a simple, streamlined workflow for maximum efficiency and productivity in your laboratory.
Use the Thermo Scientific PCR Capping Tool to quickly and easily apply caps to the SureTect lysis and PCR plates guaranteeing a tight seal in a fast and easy to use workflow
Quick and Easy
Use the Thermo Scientific PCR Capping Tools to speed the process of capping multiple lysis and assay strips. This tool is designed according to the tube and cap dimensions so you can quickly cap lysis and PCR tube strips. The rugged material and design endures constant use so can be used time and time again whatever your sample throughput.
Combined with our comprehensive portfolio of assays and world-class instrumentation you can perform food safety tests confidently knowing exactly what you are getting – quality combined with simplicity.
We are committed to enabling you to experience the true confidence in your results, right-first time, every time, and the Thermo Scientific™ SureTect™ Real-Time PCR Workflow is continually evolving to support your laboratory and testing needs.
Especificaciones
Especificaciones
| Description | Thermo Scientific™ PCR Capping Tool |
| Type | Capping Tool |
| For Use With (Equipment) | SureTect assays |
Al hacer clic en Enviar, acepta que Fisher Scientific se ponga en contacto con usted en relación con los comentarios que ha proporcionado en este formulario. No compartiremos su información para ningún otro fin. Toda la información de contacto proporcionada se mantendrá de acuerdo con nuestra Política de Privacidad. Política de privacidad.