Learn More
Thermo Scientific™ Thermo Scientific™ PCR Uncapping Tool for Lysis Tubes
Shop All Thermo Scientific ProductsDescription
Easily and quickly uncap lysis tubes in the SureTect workflow using the Thermo Scientific lysis tube uncapping tool.
Easily and quickly uncap the lysis tubes during the Thermo Scientific™ SureTect™ PCR Assay Workflow using the Thermo Scientific™ Lysis Tube Uncapping tool.
Designed specifically for use with the SureTect assays, the Thermo Scientific lysis tube uncapping tool is one in a suite of tools enabling a simple, streamlined workflow for maximum efficiency and productivity in your laboratory.
Use the Thermo Scientific Lysis Tube Uncapping tool to quickly and easily uncap the SureTect lysis tubes guaranteeing efficient cap removal in a fast and easy to use workflow
Quick and Easy
Use the Thermo Scientific Lysis Tube Uncapping tool to speed the process of uncapping multiple lysis tube strips. This tool is designed according to the tube and cap dimensions so you can quickly uncap lysis tube strips. The rugged material and design endures constant use so can be used time and time again whatever your sample throughput.
Combined with our comprehensive portfolio of assays and world-class instrumentation you can perform food safety tests confidently knowing exactly what you are getting – quality combined with simplicity.
We are committed to enabling you to experience the true confidence in your results, right-first time, every time, and the Thermo Scientific™ SureTect™ Real-Time PCR System is continually evolving to support your laboratory and testing needs.
Specifications
Specifications
| Type | Decapper |
| For Use With (Equipment) | SureTect lysis tubes |