Learn More
Thermo Scientific™ Nunc™ 96-Well Amplification Plate Accessories
Product Code. 10439192
Click to view available options
Plate Compatibility:
96-well PCR Plate
Unit Size:
Each
Product Code. 10439192
Brand: Thermo Scientific™ 259684
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
Nonsterile and autoclavable at 121°C
Certified as DNA- and RNAse-free.
  • Reusable
  • Ensures even heat transfer
  • External Dimensions: 115 × 80 × 3mm
The PCR (polymerase chain reaction) process is covered by US patents owned by Hoffmann-La Roche, Inc. and Hoffmann-La Roche Ltd.
TRUSTED_SUSTAINABILITY
Specifications
Plate Compatibility 96-well PCR Plate