Learn More
Unity Lab Services™ Thermo Scientific Liquid Nitrogen Storage Equipment Validation Service with Temperature Mapping for CryoPlus™ Storage Systems
Product Code. 15651549
Change view
Click to view available options
For Use With (Equipment):
Thermo Scientific Liquid Nitrogen Storage Equipment including Thermo Scientific CryoPlus™ Storage Systems
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. For Use With (Equipment) Unit Size
15651549 Thermo Scientific Liquid Nitrogen Storage Equipment including Thermo Scientific CryoPlus™ Storage Systems Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 15651549 Supplier Unity Lab Services™ Supplier No. IOPQPCKE89003506
Equipment and Instrument End of Year Sale
Use promo code 28000
View all promotions
 Restricted Item
Estimated Shipment: 30-12-2026
Stay compliant with our Industry-Exclusive, No-Charge Requalification Guarantee included with each Thermo Scientific Liquid Nitrogen Storage Equipment Validation Service with Temperature Mapping for Thermo Scientific CryoPlus™ Storage Systems
IQ
  • Unpacking, Assembly and Installation Verifies that the equipment, manuals, supplies and any other accessories arrived undamaged as specified in the sales order. Verifies equipment is assembled and installed and any initial diagnostics tests have been performed. Any abnormal event(s) observed during assembly and installation have been documented.
  • Utility, Facility and Environment Verifies that the installation site satisfactorily meets manufacturer-specified environmental requirements.
OQ
  • System Component Information Detailed configuration information for each system component have been recorded.
  • OQ Limits: Record a list of manufacturer recommended limits for ensuring that the system is operating as expected.
  • Equipment Operational Tests and Results: Test important equipment functions to verify that the equipment operates as intended by the manufacturer and required by the user. This is a group of selected important equipment parameters for testing according to the intended use of the equipment. - Key Tests: Door Ajar, Power Failure, Temperature Alerts
Temp Map
  • Temperature Mapping Tests and Results: Test important equipment capabilities for ensuring that the equipment is performing in accordance with the intended use of the system.
  • - Multi-Point Temperature Mapping for a 24 hour period with 1 minute data intervals. Includes Freezing Cycle testing of liquid phase cooling , super cooling, phase change to solid
TRUSTED_SUSTAINABILITY
Specifications
For Use With (Equipment) Thermo Scientific Liquid Nitrogen Storage Equipment including Thermo Scientific CryoPlus™ Storage Systems
Location Contact your local sales representative for a list of standard and extended coverage areas.
Includes Travel to major metropolitan areas. Does not include Installation. Instrument must be installed, powered-up and set temperature point for 24 hours before on-site service