Learn More
Thermo Scientific™ Hypersil™ ODS (C18) Javelin™ Guard Columns
Product Code. p-3134417 Shop All Thermo Scientific Products
Click to view available options
Diameter (Metric) Inner:
2.1 mm
4 mm
Length (Metric):
10 mm
20 mm
Particle Size:
5 μm
Unit Size:
Pack of 4
Product Code. 11533181
Brand: Thermo Scientific™ 30105024006
Only null left
Add to basket
Add to basket
Offer convenient, easy-to-use protection for analytical HPLC columns
  • Direct-connection, fingertight design for convenience and maximum efficiency
  • Patented design requires no holder, allowing quick and easy replacement
TRUSTED_SUSTAINABILITY
Specifications
Particle Size 5 μm
Length (Metric) 20 mm
Diameter (Metric) Inner 4 mm
Stationary Phase C18