Learn More
Thermo Scientific™ GENESYS™ 10 UV-Vis Spectrophotometers Cylindrical Longpath Cell Holder
Product Code. 11873754 Shop All Thermo Scientific Products
Change view
Click to view available options
Type:
Cylindrical Cell Holder
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Type Unit Size
11873754 Cylindrical Cell Holder Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 11873754 Supplier Thermo Scientific™ Supplier No. 336012000
  • ONLINE EXCLUSIVE PRICE
    3795.00 NOK
    3523.87 NOK / Pack of 1
    Save 271.13 NOK
    7% Off
Estimated Shipment: 13-01-2027
For GENESYS 10 Spectrophotometers with single-cell cuvette holder.
  • Accommodates a cylindrical cell with a 10 to 100mm pathlength, 22 to 25mm dia. and Z dimension of 11mm Models with six-position cell holders can accommodate single-cell holders using accessory Single-cell Platform and Holder (14-385-476), available separately
TRUSTED_SUSTAINABILITY
Specifications
Type Cylindrical Cell Holder
For Use With (Equipment) Genesys™ 10 UV-vis spectrophotometer
Pathlength 100 mm