Learn More
Thermo Scientific™ Wash Buffer 1 for MagJET Blood RNA Kit (concentrated)
Product Code. 13535200 Shop All Thermo Scientific Products
Change view
Click to view available options
Quantity:
80 mL
Unit Size:
80mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
13535200 80 mL 80mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 13535200 Supplier Thermo Scientific™ Supplier No. R2411
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
Wash lysed samples to remove any contaminants or inhibitors from the isolated total RNA. Thermo Scientific™ MagJET™ Wash Buffer 1 (concentrated) is a component of the MagJET™ Whole Blood RNA Kit (K2751) and may be purchased separately in 80mL aliquots.
The Thermo Scientific™ MagJET™ Whole Blood RNA Kit is designed for fast and efficient purification of total RNA from fresh or frozen whole blood treated with EDTA or citrate, blood fractions, bone marrow, or samples stabilized with PAXgene™ or Tempus™. The kit utilizes paramagnetic bead technology, enabling high yields and robust performance. High binding capacity, uniform particle size, and rapid magnetic response of MagJET magnetic beads makes the technology ideal for high throughput automatic nucleic acid purification, as well as for manual purification by low sample throughput users. The resulting high-quality RNA is free of proteins, nucleases and other contaminants or inhibitors.
TRUSTED_SUSTAINABILITY
Specifikationer
Buffer Wash Buffer
Product Line MagJET
Quantity 80 mL
For Research Use Only. Not for use in diagnostic procedures.
Produkttitel
Välj ett problem
Genom att klicka på Skicka bekräftar du att du kan bli kontaktad av Fisher Scientific angående feedbacken du har lämnat i detta formulär. Vi kommer inte att dela din information för andra ändamål. All kontaktinformation som tillhandahålls ska också underhållas i enlighet med vår Sekretesspolicy.