Learn More
Thermo Scientific™ Evolution™ One, OnePlus and Pro Spectrophotometer Solid Sample Holders
Product Code. 10061104 Shop All Thermo Scientific Products
Change view
Click to view available options
Holds:
Round/Square Optical Samples
Std 2 x 3 in. Sample Holders
Type:
Solid Sample Holder
Universal Sample Holder
Unit Size:
Each
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Holds Type Unit Size
10061104 Round/Square Optical Samples Universal Sample Holder Each
11801892 Std 2 x 3 in. Sample Holders Solid Sample Holder Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 10061104 Supplier Thermo Scientific™ Supplier No. 222217400
  • ONLINE EXCLUSIVE PRICE
    12815.00 NOK
    11913.33 NOK / Pack of 1
    Save 901.67 NOK
    7% Off
Estimated Shipment: 13-01-2027
Designed to hold solid samples for transmittance measurements
Solid Sample Holder
  • Provides easy mounting of solid samples like filters, plastic, glass and optics
  • Accommodates standard 2 x 3 in. sample holders
Universal Sample Holder
  • Holds round or square optical samples; glass or plastic samples; lenses from sunglasses or goggles
  • Requires one Solid Sample Holder (14-387-503)
TRUSTED_SUSTAINABILITY
Specifications
Type Universal Sample Holder
For Use With (Equipment) Thermo Scientific™ Evolution™ One, OnePlus and Pro UV-visible spectrophotometers
Holds Round/Square Optical Samples