Learn More
Thermo Scientific™ CryoExtra™ Racks and Accessories
Product Code. p-4331334 Shop All Thermo Scientific Products
Click to view available options
For Use With (Equipment):
Thermo Scientific CryoExtra 20, 40, 80 and 94 Rack
Thermo Scientific CryoExtra 94 Rack
Type:
Cryo Mini Rack
Cryo Square Rack
Cryo Storage Frame
Holds:
2mL, 13 Cardboard Boxes (100-cell Divider)
2mL, 13 Cardboard Boxes (25-cell Divider)
2mL, 15 Cardboard Boxes (100-cell Divider)
2mL, 15 Cardboard Boxes (25-cell Divider)
Fenwal 4R5461 (250 mL Bag), Cassettes (5)
Fenwal 4R5462 (500 mL Bag), Cassettes (5)
Fenwal 4R9951 (50 mL Bag), Cassettes (8)
Unit Size:
Each
Product Code. 15610216
Brand: Thermo Scientific™ 1950683
Only null left
Add to basket
Add to basket
Racks, frames and electrical options
TRUSTED_SUSTAINABILITY
Specifications
Type Cryo Square Rack
For Use With (Equipment) Thermo Scientific CryoExtra 94 Rack
Includes 2mL, 15 Cardboard boxes (100 cell divider)
Holds 2mL, 15 Cardboard Boxes (100-cell Divider)