Learn More
Thermo Scientific™ CryoExtra™ Racks and Accessories
Product Code. 15771267 Shop All Thermo Scientific Products
Change view
Click to view available options
For Use With (Equipment):
Thermo Scientific CryoExtra 20, 40, 80 and 94 Rack
Thermo Scientific CryoExtra 94 Rack
Type:
Cryo Mini Rack
Cryo Square Rack
Cryo Storage Frame
Holds:
2mL, 13 Cardboard Boxes (100-cell Divider)
2mL, 13 Cardboard Boxes (25-cell Divider)
2mL, 15 Cardboard Boxes (100-cell Divider)
2mL, 15 Cardboard Boxes (25-cell Divider)
Fenwal 4R5461 (250 mL Bag), Cassettes (5)
Fenwal 4R5462 (500 mL Bag), Cassettes (5)
Fenwal 4R9951 (50 mL Bag), Cassettes (8)
Unit Size:
Each
Product Code. For Use With (Equipment) Type Holds unitSize
15771267 Thermo Scientific CryoExtra 20, 40, 80 and 94 Rack Cryo Storage Frame Fenwal 4R5461 (250 mL Bag), Cassettes (5) Each
15610216 Thermo Scientific CryoExtra 94 Rack Cryo Square Rack 2mL, 15 Cardboard Boxes (100-cell Divider) Each
15152754 Thermo Scientific CryoExtra 20, 40, 80 and 94 Rack Cryo Mini Rack 2mL, 13 Cardboard Boxes (25-cell Divider) Each
15620216 Thermo Scientific CryoExtra 94 Rack Cryo Mini Rack 2mL, 15 Cardboard Boxes (25-cell Divider) Each
15761267 Thermo Scientific CryoExtra 94 Rack Cryo Storage Frame Fenwal 4R9951 (50 mL Bag), Cassettes (8) Each
15781267 Thermo Scientific CryoExtra 20, 40, 80 and 94 Rack Cryo Storage Frame Fenwal 4R5462 (500 mL Bag), Cassettes (5) Each
13449226 Thermo Scientific CryoExtra 20, 40, 80 and 94 Rack Cryo Square Rack 2mL, 13 Cardboard Boxes (100-cell Divider) Each
7 options
Product Code. 15771267
Brand: Thermo Scientific™ 1950688
Only null left
Add to basket
Add to basket
Racks, frames and electrical options
TRUSTED_SUSTAINABILITY
Specifications
Type Cryo Storage Frame
Capacity (Metric) 250 mL
For Use With (Equipment) Thermo Scientific CryoExtra 20, 40, 80 and 94 Rack
Holds Fenwal 4R5461 (250 mL Bag), Cassettes (5)