Learn More
Thermo Scientific™ Chromacol™ Storage Vial Kits with Solid Top Caps
Product Code. 10277943
Change view
Click to view available options
Color:
Clear
Unit Size:
Pack of 200
Product Code. Color unitSize
10277943 Clear Pack of 200
1 options
Product Code. 10277943
Brand: Thermo Scientific™ 8SVCP
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
Offered as amber or clear glass, in capacities from 8mL to 22mL.
TRUSTED_SUSTAINABILITY
Specifications
Color Clear
Volume (Metric) 8.0 mL