Learn More
Thermo Scientific™ BetaSil™ Silica Javelin™ Guard Columns
Product Code. 11503231 Shop All Thermo Scientific Products
Change view
Click to view available options
:
Particle Size: 5μm; 10L x 2.1mm I.D.
Product Code.
11503231 Particle Size: 5μm; 10L x 2.1mm I.D.
1 options
Product Code. 11503231
Brand: Thermo Scientific™ 70005012106
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
Economically extends HPLC column lifetime
Thermo Scientific™ BETASIL™ Silica HPLC columns are based on a highly pure, 100Å silica designed for small molecule analysis. BETASIL Silica columns are used for normal phase separations. These non-bonded silica columns are a powerful and efficient tool for separating non-polar and moderately polar compounds.
  • 100Å high purity silica
  • Used for the separation of non-polar and moderately polar compounds.
  • Direct-connection, fingertight design for convenience and maximum efficiency
  • Patented design requires no holder, allowing quick and easy replacement
Features of Betasil Silica Solid Phase
  • 100Å silica
  • High surface area with high bonded phase coverage
TRUSTED_SUSTAINABILITY
Specifications
Particle Size 5 μm
Length (Metric) 10 mm
Diameter (Metric) Inner 2.1 mm
Product Line BetaSil