Learn More
Thermo Scientific™ BetaBasic Phenyl Guard Cartridges
Product Code. 10690816 Shop All Thermo Scientific Products
Change view
Click to view available options
Diameter (Metric) Inner:
2.1 mm
3 mm
4 mm
Particle Size:
3 μm
5 μm
For Use With (Equipment):
BetaBasic™ HPLC Column
Hypersil™ HPLC Column
Unit Size:
Pack of 4
Product Code. Diameter (Metric) Inner Particle Size For Use With (Equipment) unitSize
10690816 3 mm 3 μm BetaBasic™ HPLC Column Pack of 4
10574125 2.1 mm 3 μm Hypersil™ HPLC Column Pack of 4
10340865 4 mm 5 μm BetaBasic™ HPLC Column Pack of 4
3 options
Product Code. 10690816
Brand: Thermo Scientific™ 71803011001
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
Guard columns provide convenient, easy-to-use protection for analytical HPLC columns
TRUSTED_SUSTAINABILITY
Specifications
Carbon Load 7%
Particle Size 3 μm
Length (Metric) 10 mm
Pore Size 150Å
Diameter (Metric) Inner 3 mm
For Use With (Equipment) BetaBasic™ HPLC Column
Stationary Phase Phenyl
Particle Shape Spherical
Endcapped Yes
Product Line Beta