Learn More
Thermo Scientific™ BetaBasic™ 4 DASH-HTS HPLC Columns
Shop All Thermo Scientific Products
Click to view available options
Quantity:
3/Pk.
Unit Size:
Pack of 3
Description
Thermo Scientific HPLC columns are manufactured under strict ISO 9001:2000 procedures, and every batch of silica and bonded phase is thoroughly tested for physical and chromatographic characteristics. Test reports are included with every column shipped.
- Fully deactivated Type B silica to ensure sharp peaks and excellent reproducibility
- Give less retention than C18 or C8 phases, and are a good choice for fast analyses including LC/MS of all types of compounds
- For basic compounds at high pH, which allows for a wide choice of buffer selection in LC/MS applications
- Stable bonding chemistry and excellent surface coverage gives the columns exceptional low and high pH stability
- Patented, low dead volume design minimizes band spreading
- Finger-tight fittings make changing columns quick and easy
- Available in packs of 3 or packs of 10 for better cost-effectiveness
Specifications
Specifications
| Column Format | DASH-HTS Column, 3/Pk. |
| Diameter (Metric) | 2.1 mm |
| Length (Metric) | 20 mm |
| Particle Size | 5 μm |
| Pore Size | 150 Å |
| Quantity | 3/Pk. |
| USP Type | L26 |
| Stationary Phase | C4 |
| Product Line | Beta |
| Packing Material | Silica |
| Show More |