Learn More
Thermo Scientific™ Barnstead™ GenPure™ xCAD Plus Remote Dispensers
Product Code. 15182883 Shop All Thermo Scientific Products
Click to view available options
Type:
Water Dispenser, Bench Version
Water Dispenser, Wall Version
Height (English):
25.8 in.
28.5 in.
Unit Size:
Each
Product Code. 15182883
Brand: Thermo Scientific™ 50136494
Only null left
Add to basket
Add to basket
Increase flexibility with additional remote dispensers
  • Available in bench and wall-mounted versions
  • Attach up to two additional dispensers per system
TRUSTED_SUSTAINABILITY
Specifications
For Use With (Equipment) GenPure xCAD Plus, GenPure xCAD Plus UV, GenPure xCAD Plus UF, GenPure xCAD Plus UV/UF, GenPure xCAD Plus UV - TOC, GenPure xCAD Plus UV/UF-TOC
Type Water Dispenser, Bench Version
Width (English) 10.2 in.
Width (Metric) 260 mm
Height (English) 28.5 in.
Height (Metric) 725 mm
Depth (English) 21 in.
Depth (Metric) 530 cm