Learn More
Thermo Scientific™ SoftFit-L™ Filtered Pipette Tips in Reload Inserts
GREENER_CHOICE
Product Code. p-4261872
Change view
Click to view available options
Volume (Metric):
20 μL
200 μL
300 μL
1000 μL
1200 μL
Unit Size:
Pack of 768
Pack of 960
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Volume (Metric) unitSize
12637696 1000 μL Pack of 768
11323393 200 μL Pack of 960
11303393 20 μL Pack of 960
12845063 300 μL Pack of 960
11383393 1200 μL Pack of 768
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
Product Code. 12637696 Supplier Thermo Scientific™ Supplier No. 2779RI
Only null left
Add to basket
Add to basket
Gain the accuracy and precision you expect from your pipette every time with these sterile pipette tips.
Gain the accuracy and precision you expect from your pipette samples every time with Thermo Scientific™ SoftFit-L™ pipette tips. For use with Rainin™ LTS pipettes.
  • Reload inserts are an environmentally-friendly packaging option that reduces plastic waste and storage space
  • Compatible with SoftFit-L empty racks
  • Color-coded inserts allows quick identification of tip volume
TRUSTED_SUSTAINABILITY
Specifications
Description 1000L, Filtered, Sterile, Refill
Volume (Metric) 1000 μL
Filtered Filtered
Sterility Sterile
Color Natural
Model 1000L
Length (English) 3.39 in.
Length (Metric) 8.61cm
Clean Claims DNase, RNase, DNA, ATP, and Endotoxin Free
Format Refill
Tip Style Universal
Color Code Blue
For Use With (Equipment) Rainin LTS LiteTouch
Product Line ART SoftFit L