Learn More
- Home
- Products
- Facility Safety and Maintenance
- Labels and Label Makers
- Custom Labels
- Brady™ Thermal Transfer Printable Labels
Brady™ Thermal Transfer Printable Labels
Product Code. 17100511
Change view
Click to view available options
Length (Metric):
9.53 mm
12.70 mm
25.40 mm
Color:
Blue, White
Green, White
Orange, White
Pink, White
Red, White
Yellow, White
Quantity:
10,000 Labels/Roll
2500 Labels/Roll
3000 Labels/Roll
Unit Size:
Pack of 10000
Pack of 2500
Pack of 3000
30 product options available for selection
| Product Code. | Length (Metric) | Color | Quantity | unitSize |
|---|---|---|---|---|
| 17100511 | 9.53 mm | Red, White | 3000 Labels/Roll | Pack of 3000 |
| 17190501 | 9.53 mm | Yellow, White | 3000 Labels/Roll | Pack of 3000 |
| 17110511 | 9.53 mm | Pink, White | 3000 Labels/Roll | Pack of 3000 |
| 17120511 | 9.53 mm | Orange, White | 3000 Labels/Roll | Pack of 3000 |
| 17130511 | 9.53 mm | Green, White | 3000 Labels/Roll | Pack of 3000 |
| 17140511 | 9.53 mm | Blue, White | 3000 Labels/Roll | Pack of 3000 |
| 17130501 | 12.70 mm | Yellow, White | 2500 Labels/Roll | Pack of 2500 |
| 17140501 | 12.70 mm | Red, White | 2500 Labels/Roll | Pack of 2500 |
| 17150501 | 12.70 mm | Pink, White | 2500 Labels/Roll | Pack of 2500 |
| 17160501 | 12.70 mm | Orange, White | 2500 Labels/Roll | Pack of 2500 |
| 17170501 | 12.70 mm | Green, White | 2500 Labels/Roll | Pack of 2500 |
| 17180501 | 12.70 mm | Blue, White | 2500 Labels/Roll | Pack of 2500 |
| 17150511 | 12.70 mm | Yellow, White | 3000 Labels/Roll | Pack of 3000 |
| 17160511 | 12.70 mm | Red, White | 3000 Labels/Roll | Pack of 3000 |
| 17170511 | 12.70 mm | Pink, White | 3000 Labels/Roll | Pack of 3000 |
| 17180511 | 12.70 mm | Orange, White | 3000 Labels/Roll | Pack of 3000 |
| 17190511 | 12.70 mm | Green, White | 3000 Labels/Roll | Pack of 3000 |
| 17100521 | 12.70 mm | Blue, White | 3000 Labels/Roll | Pack of 3000 |
| 17170491 | 12.70 mm | Yellow, White | 10,000 Labels/Roll | Pack of 10000 |
| 17180491 | 12.70 mm | Red, White | 10,000 Labels/Roll | Pack of 10000 |
| 17190491 | 12.70 mm | Pink, White | 10,000 Labels/Roll | Pack of 10000 |
| 17100501 | 12.70 mm | Orange, White | 10,000 Labels/Roll | Pack of 10000 |
| 17110501 | 12.70 mm | Green, White | 10,000 Labels/Roll | Pack of 10000 |
| 17120501 | 12.70 mm | Blue, White | 10,000 Labels/Roll | Pack of 10000 |
| 17110491 | 25.40 mm | Yellow, White | 3000 Labels/Roll | Pack of 3000 |
| 17120491 | 25.40 mm | Red, White | 3000 Labels/Roll | Pack of 3000 |
| 17130491 | 25.40 mm | Pink, White | 3000 Labels/Roll | Pack of 3000 |
| 17140491 | 25.40 mm | Orange, White | 3000 Labels/Roll | Pack of 3000 |
| 17150491 | 25.40 mm | Green, White | 3000 Labels/Roll | Pack of 3000 |
| 17160491 | 25.40 mm | Blue, White | 3000 Labels/Roll | Pack of 3000 |
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 30 options available.
This item is not returnable.
View return policy
This item is not returnable.
View return policy
Description
- The label will stay attached and remain legible, even when exposed to chemicals or stored in liquid nitrogen
- Thanks to subsurface printing, the label's colour code will not fade, smear or smudge and will continue to offer fast sample recognition long after it was printed
Specifications
Specifications
| Certifications/Compliance | Compliant with Directive 2011/65/EU and 2015/863 |
| Adhesive Type | Acrylic, Permanent |
| Coating | None |
| Color | Red, White |
| Finish | Gloss |
| Format | Roll |
| Material | Polyester |
| Printable Sides | Front |
| Print Technology | Ink Jet |
| Product Type | Label |
| Sterility | Non-sterile |
| For Use With (Application) | Cold temperature labelling, Component and equipment labelling, Laboratory labelling |
| For Use With (Equipment) | i5100, i7100, PR Plus, IP Series, THT 76 mm Core Printers, i7500 |
| Labels per Row | 1 |
| Min. Operating Temperature (Metric) | -196°C |
| Model | THT-304-494-3-RD |
| No. per Pack | 3000 Sets / Roll |
| Quantity | 3000 Labels/Roll |
| Shape | Rectangle with Circle |
| Size | 25.40 mm (W) x 9.53 mm (H) x 9.53 mm (Dia) |
| Temperature Range (English) | -320.8°F to +158°F |
| Temperature Range (Metric) | -196°C to +70°C |
| Thickness (English) | 0.003 in. |
| Thickness (Metric) | 0.0762 mm |
| Vertical Repeat (Metric) | 12.70 mm |
| Dimensions (LxW) | 25.40 mm (W) x 9.53 mm (H) x 9.53 mm (Dia) |
| Length (English) | 0.375 in. |
| Length (Metric) | 9.53 mm |
| Width (English) | 1 in. |
| Width (Metric) | 25.40 mm |
| Width (English) Printable Area | 25.40 mm |
| Height (English) | 0.375 in. |
| Height (Metric) | 9.53 mm |
| Diameter (English) | 0.375 in. |
| Diameter (Metric) | 9.53 mm |
| Alternate Part Number | THT-304-494-3-RD |
| Show More Show Less |