Learn More
Brady™ Thermal Transfer BBP81 Label Printer Labels, B-427 Self Laminating Vinyl
Ideal for a variety of applications. Brady Thermal Transfer BBP81 Label Printer Labels are made of vinyl cloth with repositionable rubber adhesive and can be wound inside or out with the ribbon coated side out.
Brand: Brady™ 030548
Additional Details : Weight : 1.08860kg
Description
- For use with thermal transfer printers
- Made of B-427 self laminating vinyl with permanent rubber adhesive
Specifications
UL Recognized | |
Acrylic, Permanent | |
Matte | |
Self-laminating Vinyl (B-427) | |
Wire and cable marking, panel identification | |
1 | |
Rectangle | |
50.8 x 177.8 mm (W x H) | |
0.004 in. | |
182.55 mm | |
50.80 mm | |
38.10 mm |
Suggested ribbon series: R4300; R4500; R6200 | |
Clear, White | |
IP™ Enabled | |
BBP81 Label Printer Label | |
BBP™81 Label Printer, BRADY 1244 Series, BRADY 1344 Series, BRADY 2461/3481/6441 Series, BRADY IP Series, BRADY MVP Plus Series, BRADY X Plus II Series, TAGUS™ Thermal Transfer Printer, Thermal Transfer Printers | |
500/Pk. | |
2 x 7 in. (W x H) | |
-70°C to +70°C | |
0.102 mm | |
177.80 x 50.80 mm | |
177.80 mm |