Learn More
Thermo Scientific™ Thermal Mixers and Blocks
Product Code. 15613027 Shop All Thermo Scientific Products
Change view
Click to view available options
Product Type:
Block
Mixer with Block
Holds:
20 x 0.2mL, 12 x 1.5mL Microtubes
20 x 0.5mL, 12 x 1.5mL Microtubes
20 x 2mL, 12 x 1.5mL Microtubes
24 x 1.5mL Microtubes
24 x 2.0mL Microtubes
96-well Microplates, 0.2mL Tube Strips
Unit Size:
Each
Product Code. Product Type Holds unitSize
15613027 Block 96-well Microplates, 0.2mL Tube Strips Each
15653027 Mixer with Block 20 x 0.5mL, 12 x 1.5mL Microtubes Each
15623027 Mixer with Block 24 x 1.5mL Microtubes Each
15633027 Mixer with Block 24 x 2.0mL Microtubes Each
15643027 Mixer with Block 20 x 0.2mL, 12 x 1.5mL Microtubes Each
15663027 Mixer with Block 96-well Microplates, 0.2mL Tube Strips Each
15673017 Block 24 x 1.5mL Microtubes Each
15683017 Block 24 x 2.0mL Microtubes Each
15693017 Block 20 x 2mL, 12 x 1.5mL Microtubes Each
15603027 Block 20 x 0.5mL, 12 x 1.5mL Microtubes Each
10 options
Product Code. 15613027
Brand: Thermo Scientific™ 13687716
Only null left
Add to basket
Add to basket
Get flexibility with this vertical mixer with variable speed and temperature control for a variety of applications.
Gain variable speed and temperature control with the Thermo Scientific™ Thermal Mixer, which comes with a choice of blocks for microtubes and microplates.
  • Heating and cooling capability to cover a wide range of applications
  • Compact size with quiet operation
  • Convenient packages that include a Thermal Mixer with a Block
  • Heating and cooling capability to cover a wide range of applications
  • Compact size with quiet operation
  • Convenient packages that include a Thermal Mixer with a Block
Warranty
2 years
TRUSTED_SUSTAINABILITY
Specifications
Holds 96-well Microplates, 0.2mL Tube Strips
Product Type Block
Certifications/Compliance Certificate of Analysis (COA), Certificate of Conformance (COC)