Learn More
Applied BioSystems™ Thermal Cycler Fleet Control Software
Stay in control with Thermal Cycler Fleet Control Software.
Brand: Applied BioSystems™ A40070
Additional Details : Weight : 0.00001kg
Description
Applied Biosystems™ Fleet Control Software is as intuitive and easy to use as our thermal cyclers. With this tool, you’ll be able to view and control all Applied Biosystems™ ProFlex™ PCR Systems, Automated Thermal Cyclers, and SimpliAmp™ Thermal Cyclers in your facility.
Secure—Fleet Control Software comes installed on a server that sits securely behind your firewall
Flexible—create custom permissions and rules for users, methods, and instruments
Powerful—quickly view your thermal cycler inventory and availability right from the dashboard
Convenient—control your thermal cyclers, reports, methods, and firmware upgrades from your office, another lab computer, or any other networked device
Specifications
100/240 V | |
19.5 in. Wide-Viewing Angle | |
Thermal Cycler Fleet Control Software |
Applied Biosystems™ | |
Instrument Mangement and Control | |
PCR |
For Research Use Only. Not for use in diagnostic procedures.