Learn More
- Home
- Products
- Baths
- Bath Accessories
- Julabo™ Thermal Bath Fluids
Julabo™ Thermal Bath Fluids
Product Code. 11900228
Change view
Click to view available options
Color:
Clear
Light Brown
Light Yellow
Transparent
Quantity:
5 L
10 L
Model:
H10
H20S
H250S
H350
H5
HS
HY
Thermal G
Thermal M
Temperature Range:
-20°C to +180°C
-30°C to +80°C
-40°C to +180°C
-50°C to +105°C
-80°C to +55°C
0°C to 220°C
20°C to 250°C
40°C to 170°C
50°C to 350°C
Unit Size:
10 L
5 L
Pack of 5
| Product Code. | Color | Quantity | Model | Temperature Range | unitSize |
|---|---|---|---|---|---|
| 11900228 | Transparent | 5 L | Thermal M | 40°C to 170°C | 5 L |
| 10363385 | Light Yellow | 10 L | Thermal G | -30°C to +80°C | 10 L |
| 11950278 | Clear | 5 L | H10 | -20°C to +180°C | 5 L |
| 10551802 | Light Brown | 10 L | HS | 20°C to 250°C | 10 L |
| 10048001 | Light Brown | 5 L | H20S | 0°C to 220°C | 5 L |
| 11967458 | Clear | 5 L | H5 | -50°C to +105°C | 5 L |
| 11924738 | Clear | 5 L | H350 | 50°C to 350°C | 5 L |
| 11957458 | Clear | 10 L | H10 | -40°C to +180°C | 10 L |
| 11970218 | Clear | 10 L | H5 | -50°C to +105°C | 10 L |
| 11960218 | Clear | 5 L | HY | -80°C to +55°C | 5 L |
| 11950238 | Clear | 10 L | HY | -80°C to +55°C | 10 L |
| 10162101 | Light Brown | 10 L | H20S | 0°C to 220°C | 10 L |
| 11920268 | Light Brown | 10 L | H250S | 20°C to 250°C | 10 L |
| 11989068 | Light Brown | 5 L | HS | 20°C to 250°C | 5 L |
| 11930268 | Light Brown | 5 L | H250S | 20°C to 250°C | Pack of 5 |
| 11947458 | Light Yellow | 5 L | Thermal G | -30°C to +80°C | 5 L |
| 10468881 | Transparent | 10 L | Thermal M | 40°C to 170°C | 10 L |
This item is not returnable.
View return policy
This item is not returnable.
View return policy
Description
- For closed systems like the Presto or Forte
- Has wide temperature ranges
- Low viscosity
- High stability
- Good heat conductivity
- Minimum odor
- Low corrosion tendency
- Low toxicity
- Long shelf life
Specifications
Specifications
| Color | Transparent |
| Temperature Range | 40°C to 170°C |
| Type | Thermal Bath Fluid |
| Fluid Delivery Type | Thermal Bath |
| For Use With (Application) | Ideal for temperature control applications and guarantee safe and reliable operation, viscosity, oxi |
| Includes | Drain tap |
| Model | Thermal M |
| Quantity | 5 L |
| Temperature (Metric) Maximum | >255°C |
| Viscosity | 298mm2/s at 20°C |