Learn More
Ansell ActivArmr™ 78-101 Knitted Thermal Gloves
Product Code. 11706723 Shop All Ansell Products
Change view
Click to view available options
Size:
7
9
Unit Size:
12 pairs
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Size Unit Size
11706723 9 12 pairs
12445150 7 12 pairs
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 11706723 Supplier Ansell Supplier No. 78101090
  • ONLINE EXCLUSIVE PRICE
    1168.50 NOK
    761.86 NOK / 12 pairs
    Save 406.64 NOK
    34% Off
View all promotions
Estimated Shipment: 16-12-2026
Knitted thermal work gloves feature special fibers that provide a thermal insulation barrier between the skin and surrounding environment.
  • Gloves dry faster than cotton to efficiently eliminate moisture from the skin
  • Special hollow-core fibres provide a thermal insulation barrier between the skin and surrounding environment
  • Automatic knit technology construction for longer, more comfortable wear
  • Seamless design eliminates any potential discomfort or skin irritation
  • Thermal insulation for cold weather protection
  • Ambidextrous to fit either hand
TRUSTED_SUSTAINABILITY
Specifications
ANSI Abrasion Rating ANSI 3
Certifications/Compliance (Europe) EN 420:2003 + A1:2009; Category III
Coating Material Uncoated
Color Blue
Exterior Finish No Texture
Hand Ambidextrous
Material Thermolite
Product Type Thermal Mechanic's Gloves
Size 9
EN388 Standard 214XA
For Use With (Application) Automotive Assembly, Food Processing
Glove Cuff Style Knit
Series 78-101
Yarn Gauge 13