Learn More
Abeba™ The Original 7050 Shoes
Product Code. 15662498
Click to view available options
Size:
35
36
37
38
39
40
41
42
43
44
45
46
47
Unit Size:
1 pair
Product Code. 15662498
Brand: Abeba™ 7050/38
Only null left
Add to basket
Add to basket
Animal fats, oil and fuel resistant. Sanitized™ treated.
  • CE EN ISO 20347:2012 OB A, FO SRC
  • Smooth leather
  • White
  • Slip resistance outsole sole
  • Sanitized treated
  • Leather top layer
  • adjustable instep area
Specifications
Certifications/Compliance CE EN ISO 20347:2012 OB A, FO SRC
Color White
Material Leather
Size 38
Sole Features Slip-resistant
Style 7050
Resistant To Animal fats, oil and fuel