Learn More
Cleaver Scientific The Cube Dry Bath Incubator (dual block unit),without block 220V, 1/Cs, MA: TCDB-02
Brand:  Cleaver Scientific TCDB-02
The Cube Dry Bath Incubator (dual block unit),without block 220V, 1/Cs, MA: TCDB-02
Product Code. 12398017
  • ONLINE EXCLUSIVE PRICE
    6590.00 NOK
    5897.39 NOK / Pack of 1
    Save 692.61 NOK
    10% Off
 Restricted Item
Estimated Shipment: 15-01-2027