Learn More
Thalidomide-O-amido-C3-COOH, MedChemExpress
Product Code. 16481019
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 mg
500 mg
1 g
2 g
Unit Size:
100mg
1g
2g
500mg
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16481019 1 g 1g
16464916 100 mg 100mg
16430106 2 g 2g
16411576 500 mg 500mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Product Code. 16481019 Supplier Medchem Express Supplier No. HY103612
MedChemExpress Thalidomide-O-amido-C3-COOH is a synthesized E3 ligase ligand-linker conjugate that incorporates the Thalidomide based cereblon ligand and a linker used in PROTAC technology.
Thalidomide-O-amido-C3-COOH is a synthesized E3 ligase ligand-linker conjugate that incorporates the Thalidomide based cereblon ligand and a linker used in PROTAC technology.
Specifications
Chemical Name or Material Thalidomide-O-amido-C3-COOH
CAS 2308035-51-8
Color White
Purity Grade Notes Research
Molecular Formula C19H19N3O8
Quantity 1 g
Synonym Cereblon Ligand-Linker Conjugates 7 E3 ligase Ligand-Linker Conjugates 15
Solubility Information DMSO : ≥ 55.5 mg/mL (132.98 mM)
SMILES O=C(N(C(CCC1=O)C(N1)=O)C2=O)C(C2=CC=C3)=C3OCC(NCCCC(O)=O)=O
Molecular Weight (g/mol) 417.37
Formula Weight 417.37
Percent Purity 99.49%
Grade Research
Physical Form Solid
For Use With (Application) Cancer-programmed cell death