Learn More
TH1L Rabbit anti-Human, Unconjugated, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 28417-1-AP-150UL
This item is not returnable.
View return policy
Description
The TH1L antibody from Proteintech is a rabbit polyclonal antibody generated with recombinant protein of human TH1L. This antibody recognizes human antigen. The TH1L antibody has been validated for the following applications: IF, WB, ELISA analysis.
Specifications
| TH1L | |
| Polyclonal | |
| Unconjugated | |
| 150 μL | |
| 51497 | |
| IgG |
| Western Blot, Immunofluorescence, ELISA | |
| 333 μg/mL | |
| Rabbit | |
| Primary | |
| Human |