missing translation for 'onlineSavingsMsg'
Learn More
Learn More
TH Mouse anti-Human, Mouse, Rat, Clone: 2H7B7, Proteintech
Mouse Monoclonal Antibody
Brand: Proteintech 66334-1-IG-20UL
This item is not returnable.
View return policy
Description
Tyrosine hydroxylase (TH) is an enzyme involved in the synthesis of catecholamine neurotransmitters dopamine, epinephrine, and norepinephrine. In all species, catecholamine synthesis is regulated by the interaction of TH with a cofactor, tetrahydrobiopterin (BH4). BH4 binds to the TH catalytic domain, resulting in enzymatic activity. Unlike TH in non-primate species, four human TH mRNA splice variants (hTH1-hTH4) have been isolated. These variants are identical in their catalytic domain, but differ in their N-terminal, regulatory domains. TH is also responsible for the conversion of L-tyrosine to L-dopa. TH plays a key role in the physiology of adrenergic neurons. The role of TH in the synthesis of catecholamine neurotransmitters suggests a correlation between the enzyme and a number of neuropathogenic diseases including: Parkinson′s disease, schizophrenia, Segawa syndrome, and dystonia, as well as a variety of cardiovascular diseases.Specifications
| TH | |
| Monoclonal | |
| 1 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| P04177, P07101, P24529 | |
| TH | |
| TH Fusion Protein Ag23075 | |
| 20 μL | |
| Primary | |
| Mouse, Human, Rat | |
| Antibody | |
| IgG1 |
| Immunohistochemistry (Paraffin), Western Blot, Immunofluorescence, Immunocytochemistry | |
| 2H7B7 | |
| Unconjugated | |
| TH | |
| TH, TYH, Tyrosine 3 hydroxylase, Tyrosine 3 monooxygenase, tyrosine hydroxylase | |
| Mouse | |
| Protein G | |
| RUO | |
| 21823, 25085, 7054 | |
| -20°C | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction