missing translation for 'onlineSavingsMsg'
Learn More

TGM4 Rabbit anti-Human, Mouse, Polyclonal, Proteintech

Rabbit Polyclonal Antibody

Brand:  Proteintech 15847-1-AP-20UL

Additional Details : Weight : 0.00010kg

 View more versions of this product

Product Code. 16895214

  • 1611.84 NOK / 20µL
Estimated Shipment: 22-05-2024
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

TGM4 is associated with the mammalian reproductive process. TGM4 catalyzes the cross-linking of proteins and the conjugation of polyamines to specific proteins in the seminal tract.
Specifications

Specifications

TGM4
Polyclonal
Unconjugated
TGM4
Fibrinoligase, hTGP, Prostate transglutaminase, TG(P), TGase 4, TGase P, TGM4, TGP, Transglutaminase 4, transglutaminase 4 (prostate), Transglutaminase P
Rabbit
Antigen Affinity Chromatography
RUO
331046, 7047
-20°C
Liquid
Western Blot, Immunohistochemistry (Paraffin)
0.13 mg/mL
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
P49221, Q8BZH1
Tgm4
TGM4 Fusion Protein Ag8608
20 μL
Primary
Mouse, Human
Antibody
IgG
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers