missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ TGM1 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA542057
This item is not returnable.
View return policy
Description
Peptide sequence: RGSSSGTRRP GSRGSDSRRP VSRGSGVNAA GDGTIREGML VVNGVDLLSS Sequence homology: Cow: 93%; Dog: 93%; Guinea Pig: 100%; Horse: 100%; Human: 100%; Mouse: 100%; Rabbit: 93%; Rat: 100%.
Protein kinase C (PKC) is a family of serine- and threonine-specific protein kinases that can be activated by calcium and second messenger diacylglycerol. PKC family members phosphorylate a wide variety of protein targets and are known to be involved in diverse cellular signaling pathways. PKC family members also serve as major receptors for phorbol esters, a class of tumor promoters. Each member of the PKC family has a specific expression profile and is believed to play a distinct role in cells.
Specifications
| TGM1 | |
| Polyclonal | |
| Unconjugated | |
| TGM1 | |
| 2310004J08Rik; ARCI1; Epidermal TGase; ICR2; K polypeptide; K polypeptide epidermal type I, protein-glutamine-gamma-glutamyltransferase; KTG; LI; LI1; LOW QUALITY PROTEIN: protein-glutamine gamma-glutamyltransferase K; Protein-glutamine gamma-glutamyltransferase K; protein-glutamine-gamma-glutamyltransferase; TG K; TG(K); TGASE; TGase 1; TGase K; Tgase1; TGase-1; TGK; TGM1; tranglutaminase 1; transglutaminase 1; transglutaminase 1 (K polypeptide epidermal type I, protein-glutamine-gamma-glutamyltransferase); transglutaminase 1 isoform; transglutaminase 1, K polypeptide; transglutaminase K; transglutaminase type 1; transglutaminase type I; transglutaminase, keratinocyte; transglutaminase-1 | |
| Rabbit | |
| Affinity Chromatography | |
| RUO | |
| 7051 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| Western Blot | |
| 0.5 mg/mL | |
| PBS with 2% sucrose and 0.09% sodium azide | |
| P22735 | |
| TGM1 | |
| Synthetic peptide directed towards the N-terminal of human TGM1 (aa 71-120). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction