missing translation for 'onlineSavingsMsg'
Learn More
Learn More
TGFBR3 Rabbit anti-Human, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 20000-1-AP-20UL
Additional Details : Weight : 0.00010kg
Description
This antibody is specific to TGFBR3.
This locus encodes the transforming growth factor (TGF)-beta type III receptor. The encoded receptor is a membrane proteoglycan that often functions as a co-receptor with other TGF-beta receptor superfamily members. Ectodomain shedding produces soluble TGFBR3, which may inhibit TGFB signaling. Decreased expression of this receptor has been observed in various cancers. Alternatively spliced transcript variants encoding different isoforms have been identified for this gene.Specifications
TGFBR3 | |
Polyclonal | |
Unconjugated | |
TGFBR3 | |
betaglycan, BGCAN, TGF beta receptor type 3, TGF beta receptor type III, TGF-beta Receptor III, TGFBR3, TGFR 3, TGFbeta RIII | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
7049 | |
-20°C | |
Liquid |
Flow Cytometry, Western Blot | |
0.29 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
Q03167 | |
TGFBR3 | |
Peptide | |
20 μL | |
Primary | |
Human | |
Antibody | |
IgG |