missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Description
This gene encodes a subunit of mitochondrial ATP synthase. Mitochondrial ATP synthase catalyzes ATP synthesis, utilizing an electrochemical gradient of protons across the inner membrane during oxidative phosphorylation. ATP synthase is composed of two linked multi-subunit complexes: the soluble catalytic core, F1, and the membrane-spanning component, Fo, comprising the proton channel. The catalytic portion of mitochondrial ATP synthase consists of 5 different subunits (alpha, beta, gamma, delta, and epsilon) assembled with a stoichiometry of 3 alpha, 3 beta, and a single representative of the other 3. The proton channel consists of three main subunits (a, b, c). This gene encodes the gamma subunit of the catalytic core. Alternatively spliced transcript variants encoding different isoforms have been identified. This gene also has a pseudogene on chromosome 14.
Specifications
Specifications
| Antigen | TGFBR2 |
| Applications | Western Blot |
| Classification | Monoclonal |
| Clone | 2D5H7 |
| Concentration | 0.7 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 |
| Gene | TGFBR2 |
| Gene Accession No. | P37173, P38438, Q62312 |
| Gene Alias | AAT3, FAA3, LDS1B, LDS2B, MFS2, RIIC, TAAD2, TbetaR II, TGF beta receptor type 2, TGF beta receptor type II, TGF beta type II receptor, TGFbeta RII, TGFBR2, TGFR 2 |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?