missing translation for 'onlineSavingsMsg'
Learn More
Learn More
TGF-beta 1 Antibody, R&D Systems™

Mouse Monoclonal Antibody has been used in 5 publications
Brand: R&D Systems MAB2401-100
This item is not returnable.
View return policy
Description
TGF-beta 1 Monoclonal specifically detects TGF-beta 1 in Multi-Species samples. It is validated for Western Blot.
Specifications
| TGF-beta 1 | |
| Monoclonal | |
| Unconjugated | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
| TGFB1 | |
| Chinese hamster ovary cell line CHO-derived recombinant human TGF-beta 1 | |
| 100 μg | |
| Primary | |
| Detects multiple species of TGF-beta 1 in direct ELISAs and Western blots. In direct ELISAs, this antibody does not cross-react with recombinant human (rh) TGF-alpha , rhTGF-beta 2, rhTGF-beta 3, rhTGF-beta 5, rhLAP (TGF-beta 1), rhART, rhNeurturin, or recombinant mouse TRANCE. | |
| Multi-species | |
| Supernatant |
| Western Blot | |
| 141322 | |
| Western Blot 1 ug/mL | |
| CEDLAP, DPD1, latency-associated peptide, TGF beta, TGF beta1, TGFB, TGFB1, TGFbeta, TGFbeta 1, TGF-beta 1 protein, TGF-beta-1, transforming growth factor beta-1, transforming growth factor, beta 1 | |
| Mouse | |
| Protein A or G purified from hybridoma culture supernatant | |
| RUO | |
| 7040 | |
| Reconstitute at 0.5 mg/mL in sterile PBS. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. | |
| IgG2b |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction