Learn More
TGF-beta 1 Antibody, R&D Systems™
GREENER_CHOICE
Mouse Monoclonal Antibody has been used in 5 publications
Brand:  R&D Systems MAB2401-500
 View more versions of this product
Product Code. 18767799
  • 20305.00 NOK / 500µg
Description
TGF-beta 1 Monoclonal specifically detects TGF-beta 1 in Multi-Species samples. It is validated for Western Blot.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
TGF-beta 1
Monoclonal
Unconjugated
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative
TGFB1
Chinese hamster ovary cell line CHO-derived recombinant human TGF-beta 1
500 μg
Primary
Detects multiple species of TGF-beta 1 in direct ELISAs and Western blots. In direct ELISAs, this antibody does not cross-react with recombinant human (rh) TGF-alpha , rhTGF-beta 2, rhTGF-beta 3, rhTGF-beta 5, rhLAP (TGF-beta 1), rhART, rhNeurturin, or recombinant mouse TRANCE.
Multi-species
Supernatant
Western Blot
141322
Western Blot 1 ug/mL
CEDLAP, DPD1, latency-associated peptide, TGF beta, TGF beta1, TGFB, TGFB1, TGFbeta, TGFbeta 1, TGF-beta 1 protein, TGF-beta-1, transforming growth factor beta-1, transforming growth factor, beta 1
Mouse
Protein A or G purified from hybridoma culture supernatant
RUO
7040
Reconstitute at 0.5 mg/mL in sterile PBS.
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
IgG2b